Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
AGCCATTGATAGTAGGAGTGTGTTA[C/T]ATCCTTCAGGTATGTAGGGCTGAAC
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
10 submissions
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 609238 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
LOC102724601 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
LOC102724601 - uncharacterized LOC102724601 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
RABGAP1L - RAB GTPase activating protein 1 like | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001035230.2 | Intron | NP_001030307.1 | ||||
NM_001243763.1 | Intron | NP_001230692.1 | ||||
NM_001243764.1 | Intron | NP_001230693.1 | ||||
NM_001243765.1 | Intron | NP_001230694.1 | ||||
NM_014857.4 | Intron | NP_055672.3 | ||||
XM_005245680.1 | Intron | XP_005245737.1 | ||||
XM_005245681.2 | Intron | XP_005245738.1 | ||||
XM_006711693.1 | Intron | XP_006711756.1 | ||||
XM_011510223.2 | Intron | XP_011508525.1 | ||||
XM_017002994.1 | Intron | XP_016858483.1 | ||||
XM_017002995.1 | Intron | XP_016858484.1 | ||||
XM_017002996.1 | Intron | XP_016858485.1 | ||||
XM_017002997.1 | Intron | XP_016858486.1 | ||||
XM_017002998.1 | Intron | XP_016858487.1 | ||||
XM_017002999.1 | Intron | XP_016858488.1 |