Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
AGGCCACCAGCACAGCCAGTATGGT[G/T]TCATTCACAGCGGTGTCAGCACACG
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
17 submissions
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 606416 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
NLRP3 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
NLRP3 - NLR family pyrin domain containing 3 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
OR2B11 - olfactory receptor family 2 subfamily B member 11 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001004492.1 | 600 | Missense Mutation | GAA,GAC | E,D 200 | NP_001004492.1 |