Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
ATAAAATGGACAACCACCATTATTA[G/T]TAGCTCATCTCTTAAGGTAAAGGGG
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
2 submissions
|
||||||||||||||||||||
Phenotype: |
|||||||||||||||||||||
Literature Links: |
C1orf146 PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
C1orf146 - chromosome 1 open reading frame 146 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001012425.1 | 317 | Missense Mutation | AGT,ATT | S,I 17 | NP_001012425.1 | |
XM_011541447.2 | 317 | Missense Mutation | AGT,ATT | S,I 17 | XP_011539749.1 |