Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
AATCTTTTTCCTCCAGGATGTTCCT[G/T]CAGGAGGAAGCAGCACTAAAAGCAC
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
1 submissions
|
||||||||||||||||||||
Phenotype: |
MIM: 602672 MIM: 603702 | ||||||||||||||||||||
Literature Links: |
NUP210L PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
NUP210L - nucleoporin 210 like | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001159484.1 | 270 | Intron | NP_001152956.1 | |||
NM_207308.2 | 270 | Intron | NP_997191.2 | |||
XM_011510122.1 | 270 | Intron | XP_011508424.1 | |||
XM_011510123.1 | 270 | Intron | XP_011508425.1 | |||
XM_011510124.1 | 270 | Intron | XP_011508426.1 | |||
XM_017002788.1 | 270 | Intron | XP_016858277.1 | |||
XM_017002789.1 | 270 | Intron | XP_016858278.1 |
RAB13 - RAB13, member RAS oncogene family | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
RPS27 - ribosomal protein S27 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001030.4 | 270 | Missense Mutation | NP_001021.1 |