Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
CAGCATCTCCAGAGTAGTCAAGACA[A/G]TTTGCATTCCACATGCCAATGCCCC
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
18 submissions
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 600873 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
CTBS PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
CTBS - chitobiase | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_004388.2 | 1136 | Silent Mutation | NP_004379.1 |
SPATA1 - spermatogenesis associated 1 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001310156.1 | 1136 | Intron | NP_001297085.1 | |||
XM_011542514.2 | 1136 | Intron | XP_011540816.1 | |||
XM_011542515.2 | 1136 | Intron | XP_011540817.1 | |||
XM_011542516.1 | 1136 | Intron | XP_011540818.1 | |||
XM_011542517.1 | 1136 | Intron | XP_011540819.1 | |||
XM_011542519.2 | 1136 | Intron | XP_011540821.1 | |||
XM_011542520.2 | 1136 | Intron | XP_011540822.1 | |||
XM_011542521.2 | 1136 | Intron | XP_011540823.1 | |||
XM_011542522.2 | 1136 | Intron | XP_011540824.1 |