Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
CTAGCTGTGTGGCATTGTGCTAGAC[A/G]TAATCACTCAAACCTGTGCGCTGTC
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
9 submissions
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 604345 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
MAN1A2 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
MAN1A2 - mannosidase alpha class 1A member 2 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_006699.3 | Intron | NP_006690.1 | ||||
XM_006710302.2 | Intron | XP_006710365.1 | ||||
XM_011540536.2 | Intron | XP_011538838.1 | ||||
XM_017000115.1 | Intron | XP_016855604.1 |