Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
CCGAGCTCTACAGCAGCCCGAGCGC[C/T]GCCTGCATCCAGTGCCTGGACTGCC
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
4 submissions
|
||||||||||||||||||||
Phenotype: |
MIM: 164780 | ||||||||||||||||||||
Literature Links: |
SKI PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
SKI - SKI proto-oncogene | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_003036.3 | 768 | Silent Mutation | GCC,GCT | A,A 242 | NP_003027.1 | |
XM_005244775.3 | 768 | Silent Mutation | GCC,GCT | A,A 242 | XP_005244832.1 | |
XM_005244776.4 | 768 | Intron | XP_005244833.1 | |||
XM_017002128.1 | 768 | Intron | XP_016857617.1 |