Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
AGCGCCGGGTATCCATGTAATCTGG[C/G]GGTGCAGCAGGGCTGGGCTCCTCAA
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
6 submissions
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 609738 MIM: 604634 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
IGSF9 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
IGSF9 - immunoglobulin superfamily member 9 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001135050.1 | 3054 | Silent Mutation | CCC,CCG | P,P 952 | NP_001128522.1 | |
NM_020789.3 | 3054 | Silent Mutation | CCC,CCG | P,P 936 | NP_065840.2 | |
XM_017001884.1 | 3054 | Intron | XP_016857373.1 |
TAGLN2 - transgelin 2 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |