Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
AGCTGGAATGGTGAGACCAACATCC[A/G]TGACACCTGGACTCTTTCAGGTTCT
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
2 submissions
|
||||||||||||||||||||
Phenotype: |
MIM: 610816 | ||||||||||||||||||||
Literature Links: |
SLC25A33 PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
SLC25A33 - solute carrier family 25 member 33 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_032315.2 | 218 | Intron | NP_115691.1 | |||
XM_005263503.3 | 218 | Intron | XP_005263560.1 | |||
XM_011542296.1 | 218 | Missense Mutation | ATG,GTG | M,V 7 | XP_011540598.1 |