Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
GAGCGGTCCATTACGCAGATTAAGT[A/G]TAACCGCGAAGGAGACCTCCTCTTT
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
7 submissions
|
||||||||||||||||||||
Phenotype: |
MIM: 603911 | ||||||||||||||||||||
Literature Links: |
DCDC2B PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
DCDC2B - doublecortin domain containing 2B | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
EIF3I - eukaryotic translation initiation factor 3 subunit I | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_003757.3 | 586 | Missense Mutation | TAT,TGT | Y,C 18 | NP_003748.1 | |
XM_017002671.1 | 586 | UTR 5 | XP_016858160.1 |
MTMR9LP - myotubularin related protein 9-like, pseudogene | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |