Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
CCCATGACTGATATGTTTCAGTTGC[A/G]CACCTGTTTGTCAGAAAAGGGATTG
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
9 submissions
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 608854 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
OSCP1 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
OSCP1 - organic solute carrier partner 1 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_145047.4 | Intron | NP_659484.4 | ||||
NM_206837.2 | Intron | NP_996668.1 | ||||
XM_005270461.1 | Intron | XP_005270518.1 | ||||
XM_005270462.1 | Intron | XP_005270519.1 | ||||
XM_005270463.3 | Intron | XP_005270520.1 | ||||
XM_011540680.2 | Intron | XP_011538982.1 | ||||
XM_011540681.2 | Intron | XP_011538983.1 | ||||
XM_017000303.1 | Intron | XP_016855792.1 | ||||
XM_017000304.1 | Intron | XP_016855793.1 |