Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
CTTGGCCTCCATCAGGAGCTGTCTG[A/G]AAGAGGGGCTGGAGCCCACCTGCTT
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
2 submissions
|
||||||||||||||||||||
Phenotype: |
MIM: 136132 | ||||||||||||||||||||
Literature Links: |
FMO3 PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
FMO3 - flavin containing monooxygenase 3 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001002294.2 | 178 | Missense Mutation | AAA,GAA | K,E 23 | NP_001002294.1 | |
NM_001319173.1 | 178 | UTR 5 | NP_001306102.1 | |||
NM_001319174.1 | 178 | Missense Mutation | AAA,GAA | K,E 23 | NP_001306103.1 | |
NM_006894.5 | 178 | Missense Mutation | AAA,GAA | K,E 23 | NP_008825.4 | |
XM_011509345.2 | 178 | UTR 5 | XP_011507647.1 |
MIR1295A - microRNA 1295a | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
MIR1295B - microRNA 1295b | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |