Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
ATCGCTCTGCGTGTATCTGTACATC[A/T]GGAGAATACGGGATTAAGTTCTCCT
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
2 submissions
|
||||||||||||||||||||
Phenotype: |
MIM: 616846 MIM: 609890 | ||||||||||||||||||||
Literature Links: |
EMC1 PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
EMC1 - ER membrane protein complex subunit 1 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001271427.1 | 2750 | Silent Mutation | CCA,CCT | P,P 899 | NP_001258356.1 | |
NM_001271428.1 | 2750 | Silent Mutation | CCA,CCT | P,P 899 | NP_001258357.1 | |
NM_001271429.1 | 2750 | Silent Mutation | CCA,CCT | P,P 878 | NP_001258358.1 | |
NM_015047.2 | 2750 | Silent Mutation | CCA,CCT | P,P 900 | NP_055862.1 | |
XM_005245787.1 | 2750 | Silent Mutation | CCA,CCT | P,P 903 | XP_005245844.1 | |
XM_005245788.1 | 2750 | Silent Mutation | CCA,CCT | P,P 902 | XP_005245845.1 |
LOC101927895 - uncharacterized LOC101927895 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
UBR4 - ubiquitin protein ligase E3 component n-recognin 4 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |