Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
CAGCAACACAAGTGACCTGAGCAGC[A/G]TGAGCTATAAAGGAGAGAGAAAGAG
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
5 submissions
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 603270 MIM: 611734 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
ATP5F1 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
ATP5F1 - ATP synthase, H+ transporting, mitochondrial Fo complex subunit B1 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
WDR77 - WD repeat domain 77 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001317062.1 | 637 | Silent Mutation | CAC,CAT | H,H 166 | NP_001303991.1 | |
NM_001317063.1 | 637 | Silent Mutation | CAC,CAT | H,H 166 | NP_001303992.1 | |
NM_001317064.1 | 637 | Silent Mutation | CAC,CAT | H,H 102 | NP_001303993.1 | |
NM_024102.3 | 637 | Silent Mutation | CAC,CAT | H,H 166 | NP_077007.1 |