Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
GATTTGTTTTTCAGAAACAAAAACG[A/G]AGATCCGTCAACTCCAAAATTCCTG
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
11 submissions
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 604538 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
KIF2C PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
KIF2C - kinesin family member 2C | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001297655.1 | 508 | Silent Mutation | CGA,CGG | R,R 93 | NP_001284584.1 | |
NM_001297656.1 | 508 | Silent Mutation | CGA,CGG | R,R 39 | NP_001284585.1 | |
NM_001297657.1 | 508 | UTR 5 | NP_001284586.1 | |||
NM_006845.3 | 508 | Silent Mutation | CGA,CGG | R,R 93 | NP_006836.2 | |
XM_011540540.2 | 508 | UTR 5 | XP_011538842.1 | |||
XM_011540541.2 | 508 | UTR 5 | XP_011538843.1 |