Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
GGGGAATGATGGCCACAGTGTAGCA[A/G]GTCTCAGAGATGGATAGCATGCACA
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
2 submissions
|
||||||||||||||||||||
Phenotype: |
MIM: 147140 | ||||||||||||||||||||
Literature Links: |
FCER1A PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
FCER1A - Fc fragment of IgE receptor Ia | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
OR10J3 - olfactory receptor family 10 subfamily J member 3 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001004467.1 | 216 | Silent Mutation | ACC,ACT | T,T 72 | NP_001004467.1 |