Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
GACGTGCTGTTTGCATCGGGGCCCC[A/G]GGAGCTCCTGAAGAAGTTCCGGCAG
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
3 submissions
|
||||||||||||||||||||
Phenotype: |
MIM: 153454 | ||||||||||||||||||||
Literature Links: |
PLOD1 PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
PLOD1 - procollagen-lysine,2-oxoglutarate 5-dioxygenase 1 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_000302.3 | 586 | Missense Mutation | CAG,CGG | Q,R 111 | NP_000293.2 | |
NM_001316320.1 | 586 | Missense Mutation | CAG,CGG | Q,R 158 | NP_001303249.1 |