Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
ATCAAAATTGACAACCAGCCTGACA[C/T]TGATCAAGTCTAGGCCTCGTCCCAA
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
20 submissions
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 615464 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
DDX59 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
DDX59 - DEAD-box helicase 59 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001031725.5 | 1954 | Missense Mutation | AAT,AGT | N,S 513 | NP_001026895.2 | |
NM_001320181.1 | 1954 | Missense Mutation | AAT,AGT | N,S 513 | NP_001307110.1 | |
NM_001320182.1 | 1954 | Missense Mutation | AAT,AGT | N,S 399 | NP_001307111.1 | |
XM_005245519.1 | 1954 | Intron | XP_005245576.1 | |||
XM_005245521.2 | 1954 | Missense Mutation | AAT,AGT | N,S 429 | XP_005245578.1 | |
XM_006711562.1 | 1954 | Missense Mutation | AAT,AGT | N,S 513 | XP_006711625.1 | |
XM_011510035.2 | 1954 | Missense Mutation | AAT,AGT | N,S 513 | XP_011508337.1 | |
XM_017002431.1 | 1954 | Intron | XP_016857920.1 | |||
XM_017002432.1 | 1954 | Missense Mutation | AAT,AGT | N,S 513 | XP_016857921.1 |