Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TCAGCACTTCATCTGTAATCTTCTG[C/T]ATCAGGAAGTTCTGTGTTTTTTCTC
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
2 submissions
|
||||||||||||||||||||
Phenotype: |
MIM: 603517 | ||||||||||||||||||||
Literature Links: |
BCL10 PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
BCL10 - B-cell CLL/lymphoma 10 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001320715.1 | 709 | Missense Mutation | ATA,ATG | I,M 96 | NP_001307644.1 | |
NM_003921.4 | 709 | Missense Mutation | ATA,ATG | I,M 96 | NP_003912.1 | |
XM_011542397.2 | 709 | Missense Mutation | ATA,ATG | I,M 149 | XP_011540699.1 | |
XM_011542398.2 | 709 | Missense Mutation | ATA,ATG | I,M 149 | XP_011540700.1 | |
XM_011542399.2 | 709 | Missense Mutation | ATA,ATG | I,M 78 | XP_011540701.1 |
LOC646626 - uncharacterized LOC646626 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |