Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TACCATGTGGTGGCTGCTTTTCCCC[C/G]ACCAGGGCCTCAGCTCCAGCAGCTG
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
5 submissions
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 142622 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
HPCA PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
HPCA - hippocalcin | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_002143.2 | 769 | Intron | NP_002134.2 | |||
XM_005270792.2 | 769 | Intron | XP_005270849.1 | |||
XM_017001118.1 | 769 | Intron | XP_016856607.1 |
TMEM54 - transmembrane protein 54 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
XM_005270408.3 | 769 | Missense Mutation | TCG,TGG | S,W 191 | XP_005270465.1 |