Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
AAAAAACGAGCAGAATTTTAAACTT[A/G]CAGGTACGTGTTTAAATCCAAAGAT
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
12 submissions
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 603311 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
CDC7 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
CDC7 - cell division cycle 7 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001134419.1 | 277 | Missense Mutation | ACA,GCA | T,A 38 | NP_001127891.1 | |
NM_001134420.1 | 277 | Missense Mutation | ACA,GCA | T,A 38 | NP_001127892.1 | |
NM_003503.3 | 277 | Missense Mutation | ACA,GCA | T,A 38 | NP_003494.1 | |
XM_005271241.2 | 277 | Missense Mutation | ACA,GCA | T,A 38 | XP_005271298.1 | |
XM_005271244.2 | 277 | Missense Mutation | ACA,GCA | T,A 38 | XP_005271301.1 | |
XM_017002425.1 | 277 | Missense Mutation | ACA,GCA | T,A 38 | XP_016857914.1 | |
XM_017002426.1 | 277 | Missense Mutation | ACA,GCA | T,A 38 | XP_016857915.1 | |
XM_017002427.1 | 277 | Intron | XP_016857916.1 |