Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
GGATAGCTGGGGAGTTGTCAGGCAG[G/T]AACATTGATGTAGTCATCTGAATCA
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
21 submissions
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 606015 MIM: 604136 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
FCMR PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
FCMR - Fc fragment of IgM receptor | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001142473.1 | 1285 | Missense Mutation | ACT,CCT | T,P 277 | NP_001135945.1 | |
NM_001193338.1 | 1285 | UTR 3 | NP_001180267.1 | |||
NM_005449.4 | 1285 | Missense Mutation | ACT,CCT | T,P 389 | NP_005440.1 | |
XM_005273351.3 | 1285 | UTR 3 | XP_005273408.1 | |||
XM_005273352.4 | 1285 | Missense Mutation | ACT,CCT | T,P 298 | XP_005273409.1 |
IL24 - interleukin 24 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001185156.1 | 1285 | Intron | NP_001172085.1 | |||
NM_001185157.1 | 1285 | Intron | NP_001172086.1 | |||
NM_001185158.1 | 1285 | Intron | NP_001172087.1 | |||
NM_006850.3 | 1285 | Intron | NP_006841.1 | |||
XM_011509101.2 | 1285 | Intron | XP_011507403.1 | |||
XM_017000121.1 | 1285 | Intron | XP_016855610.1 |