Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
ACAGTGACGTACCGTTGCCAGTCCT[A/T]CTATAAACTCCAGGGCTCTGTAACT
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
1 submissions
|
||||||||||||||||||||
Phenotype: |
MIM: 608593 | ||||||||||||||||||||
Literature Links: |
CFHR5 PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
CFHR5 - complement factor H related 5 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_030787.3 | 1570 | Missense Mutation | TAC,TTC | Y,F 481 | NP_110414.1 | |
XM_011510020.2 | 1570 | Missense Mutation | TAC,TTC | Y,F 484 | XP_011508322.1 |