Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
ACAACCCTTTACAAGGCCAGAAATG[C/T]ATTGTTCAAACAACTTCATGGTCCC
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
9 submissions
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 602369 MIM: 604743 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
CYR61 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
CYR61 - cysteine rich angiogenic inducer 61 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001554.4 | 911 | Silent Mutation | TGC,TGT | C,C 229 | NP_001545.2 |
DDAH1 - dimethylarginine dimethylaminohydrolase 1 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |