Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
CAGATTAAAAATGCGTTCTGTTGAC[A/G]TAACATTGGTCCTCCTGAGTTTTGT
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
2 submissions
|
||||||||||||||||||||
Phenotype: |
MIM: 611146 | ||||||||||||||||||||
Literature Links: |
SLC30A10 PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
SLC30A10 - solute carrier family 30 member 10 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_018713.2 | 1984 | Silent Mutation | TAC,TAT | Y,Y 479 | NP_061183.2 | |
XM_006711437.3 | 1984 | Silent Mutation | TAC,TAT | Y,Y 416 | XP_006711500.2 | |
XM_017001684.1 | 1984 | Silent Mutation | TAC,TAT | Y,Y 320 | XP_016857173.1 |