Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TGGTGCTCAACAGTGGGGAGAAAGG[C/T]GGTGACTCCTTTGGGGTCCAAATCA
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
7 submissions
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 609473 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
CGN PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
CGN - cingulin | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_020770.2 | 366 | Silent Mutation | GGC,GGT | G,G 74 | NP_065821.1 | |
XM_005245365.4 | 366 | Silent Mutation | GGC,GGT | G,G 74 | XP_005245422.1 |