Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
AGCCCCGGCGGATCATCCTCCTGGT[A/G]AAGGCTGGGCAAGCTGTGGATGATT
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
4 submissions
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 172200 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
PGD PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
PGD - phosphogluconate dehydrogenase | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001304451.1 | 351 | Silent Mutation | GTA,GTG | V,V 75 | NP_001291380.1 | |
NM_001304452.1 | 351 | Silent Mutation | GTA,GTG | V,V 62 | NP_001291381.1 | |
NM_002631.3 | 351 | Silent Mutation | GTA,GTG | V,V 75 | NP_002622.2 |