Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
AGCCCAGCCTGGAAGCACGGGTGCG[C/T]GACATCGCCATAGCAACAAGGAACA
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
5 submissions
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 612316 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
ATAD3A PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
ATAD3A - ATPase family, AAA domain containing 3A | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001170535.1 | 1257 | Silent Mutation | CGC,CGT | R,R 329 | NP_001164006.1 | |
NM_001170536.1 | 1257 | Silent Mutation | CGC,CGT | R,R 250 | NP_001164007.1 | |
NM_018188.3 | 1257 | Silent Mutation | CGC,CGT | R,R 377 | NP_060658.3 | |
XM_017001628.1 | 1257 | Silent Mutation | CGC,CGT | R,R 329 | XP_016857117.1 |