Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
ACAGGACTTCCTGGCAGGACAAGGG[C/G]AGCTCCAGGCCCAAGGCTCCCGTGG
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
1 submissions
|
||||||||||||||||||||
Phenotype: |
MIM: 605951 MIM: 191164 MIM: 613683 | ||||||||||||||||||||
Literature Links: |
DPM3 PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
DPM3 - dolichyl-phosphate mannosyltransferase subunit 3 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_018973.3 | 125 | Silent Mutation | CTC,CTG | L,L 60 | NP_061846.2 | |
NM_153741.1 | 125 | Silent Mutation | CTC,CTG | L,L 30 | NP_714963.1 | |
XM_017001498.1 | 125 | Silent Mutation | CTC,CTG | L,L 30 | XP_016856987.1 |
EFNA1 - ephrin A1 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
SLC50A1 - solute carrier family 50 member 1 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |