Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
CCCCTCCCTTCTCCAGTGTCCACCC[C/T]GCTTGGCCAAGGCCGGGTCAATCAG
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
3 submissions
|
||||||||||||||||||||
Phenotype: |
MIM: 167410 | ||||||||||||||||||||
Literature Links: |
PAX7 PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
PAX7 - paired box 7 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001135254.1 | 693 | Missense Mutation | CCG,CTG | P,L 32 | NP_001128726.1 | |
NM_002584.2 | 693 | Missense Mutation | CCG,CTG | P,L 32 | NP_002575.1 | |
NM_013945.2 | 693 | Missense Mutation | CCG,CTG | P,L 32 | NP_039236.1 |