Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
CCTCTGTCTCCAGGAAGCAGAGGCA[A/G]AAGTGATCCTTGCTGAGGAGGGCCA
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
3 submissions
|
||||||||||||||||||||
Phenotype: |
MIM: 604266 MIM: 611460 MIM: 606040 | ||||||||||||||||||||
Literature Links: |
MEGF6 PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
MEGF6 - multiple EGF like domains 6 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
TPRG1L - tumor protein p63 regulated 1-like | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_182752.3 | 1396 | Intron | NP_877429.2 |
WRAP73 - WD repeat containing, antisense to TP73 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_017818.3 | 1396 | Silent Mutation | TTC,TTT | F,F 438 | NP_060288.3 | |
XM_005244754.1 | 1396 | Silent Mutation | TTC,TTT | F,F 393 | XP_005244811.1 | |
XM_017001387.1 | 1396 | Silent Mutation | TTC,TTT | F,F 431 | XP_016856876.1 |