Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
GACAGGGCAGAGGTGGGTGGGGTGG[A/G]GTGAGCAGGAGGCTTGTCCATTGCG
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
6 submissions
|
||||||||||||||||||||
Phenotype: |
MIM: 608481 | ||||||||||||||||||||
Literature Links: |
SLC26A9 PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
SLC26A9 - solute carrier family 26 member 9 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_052934.3 | 2596 | UTR 3 | NP_443166.1 | |||
NM_134325.2 | 2596 | Missense Mutation | CCC,TCC | P,S 828 | NP_599152.2 | |
XM_011509121.2 | 2596 | UTR 3 | XP_011507423.1 | |||
XM_011509122.2 | 2596 | UTR 3 | XP_011507424.1 | |||
XM_011509124.2 | 2596 | Intron | XP_011507426.1 |