Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
AATGGCTGTGTCCAAAACTGGTGGA[A/T]AAGTGTCGCTGAAACGCAAGTACAT
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
11 submissions
|
||||||||||||||||||||
Phenotype: |
|||||||||||||||||||||
Literature Links: |
OR6K3 PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
OR6K3 - olfactory receptor family 6 subfamily K member 3 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001005327.2 | 794 | Missense Mutation | TAT,TTT | Y,F 265 | NP_001005327.2 |