Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TTCCAGCAACTTCCTCTTGTCTGAT[G/T]GCTGTGAATATTTCAAGCACAAGAG
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
10 submissions
|
||||||||||||||||||||
Phenotype: |
MIM: 609605 | ||||||||||||||||||||
Literature Links: |
MAP1LC3C PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
MAP1LC3C - microtubule associated protein 1 light chain 3 gamma | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001004343.2 | Intron | NP_001004343.1 | ||||
XM_005273139.3 | Intron | XP_005273196.1 |