Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TCCCTCTGGCTGGATTCAGACTTGG[C/T]ATGTTCCCGCTCCTTTGACACGGAG
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
3 submissions
|
||||||||||||||||||||
Phenotype: |
MIM: 601940 | ||||||||||||||||||||
Literature Links: |
SRSF4 PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
SRSF4 - serine and arginine rich splicing factor 4 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_005626.4 | 1335 | Missense Mutation | ACC,GCC | T,A 418 | NP_005617.2 | |
XM_011541951.2 | 1335 | Missense Mutation | ACC,GCC | T,A 388 | XP_011540253.1 |