Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
AGATGTGTGACGGGAGCCACTTGGC[C/T]TCCACCCTCCGCTATTGCATGACAG
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
5 submissions
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
|||||||||||||||||||||||||||||||||||||||
Literature Links: |
TMEM61 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
TMEM61 - transmembrane protein 61 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_182532.2 | 441 | Silent Mutation | GCC,GCT | A,A 13 | NP_872338.1 | |
XM_005270586.4 | 441 | Silent Mutation | GCC,GCT | A,A 8 | XP_005270643.1 | |
XM_011540911.2 | 441 | Silent Mutation | GCC,GCT | A,A 13 | XP_011539213.1 | |
XM_011540912.2 | 441 | Silent Mutation | GCC,GCT | A,A 8 | XP_011539214.1 |