Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
AATATGAGTTTGACATCGAGTTTGA[C/T]GTGAGTGTGATAGAGTGGGAAATAT
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
16 submissions
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 600923 MIM: 610554 MIM: 604729 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
PPOX PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
PPOX - protoporphyrinogen oxidase | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
UFC1 - ubiquitin-fold modifier conjugating enzyme 1 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_016406.3 | 618 | Silent Mutation | GAC,GAT | D,D 85 | NP_057490.2 | |
XM_005245254.1 | 618 | Silent Mutation | GAC,GAT | D,D 85 | XP_005245311.1 | |
XM_017001450.1 | 618 | Silent Mutation | GAC,GAT | D,D 85 | XP_016856939.1 | |
XM_017001451.1 | 618 | Silent Mutation | GAC,GAT | D,D 85 | XP_016856940.1 |
USP21 - ubiquitin specific peptidase 21 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |