Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
CATTTGACTGTGGTGGCAAAGGGAC[A/G]GCACTGAGCATGCCTAACCTATTCC
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
5 submissions
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
|||||||||||||||||||||||||||||||||||||||
Literature Links: |
YIPF1 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
YIPF1 - Yip1 domain family member 1 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_018982.4 | 1332 | UTR 3 | NP_061855.1 | |||
XM_017001499.1 | 1332 | UTR 3 | XP_016856988.1 | |||
XM_017001500.1 | 1332 | UTR 3 | XP_016856989.1 | |||
XM_017001501.1 | 1332 | UTR 3 | XP_016856990.1 |