Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
CAGAGGAACCAGCGACTTCATCCAG[C/G]AGTCTTCAGCCAGGACTACCCGGAA
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
2 submissions
|
||||||||||||||||||||
Phenotype: |
|||||||||||||||||||||
Literature Links: |
AGMAT PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
AGMAT - agmatinase | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_024758.4 | 892 | Missense Mutation | TCC,TGC | S,C 250 | NP_079034.3 |
DNAJC16 - DnaJ heat shock protein family (Hsp40) member C16 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |