Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TACTGAGTCTACGCAGTTGCCTGCT[A/G]ATTTCGGTAGCATCACTCAATTTCT
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
6 submissions
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 610274 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
PIGV PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
PIGV - phosphatidylinositol glycan anchor biosynthesis class V | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001202554.1 | 1037 | Silent Mutation | CTA,CTG | L,L 116 | NP_001189483.1 | |
NM_017837.3 | 1037 | Silent Mutation | CTA,CTG | L,L 116 | NP_060307.2 |