Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TAGGCTGAAGACGCAGAGGGGAGCG[C/T]TGTGGGTTTGGAAGATGTATGTAAT
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
17 submissions
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 609321 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
MFSD14A PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
MFSD14A - major facilitator superfamily domain containing 14A | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
SASS6 - SAS-6 centriolar assembly protein | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001304829.1 | 2122 | Missense Mutation | ACG,GCG | T,A 474 | NP_001291758.1 | |
NM_194292.2 | 2122 | Missense Mutation | ACG,GCG | T,A 641 | NP_919268.1 | |
XM_017000486.1 | 2122 | Missense Mutation | ACG,GCG | T,A 614 | XP_016855975.1 |