Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TCCTCCTCCAGGAACACTTTGCTCA[C/G]GTAGCAGGCGAACACGAAGCCGAAC
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
17 submissions
|
||||||||||||||||||||
Phenotype: |
MIM: 612871 | ||||||||||||||||||||
Literature Links: |
NKAIN1 PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
NKAIN1 - Na+/K+ transporting ATPase interacting 1 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_024522.2 | 839 | Missense Mutation | CTG,GTG | L,V 167 | NP_078798.2 | |
XM_017002320.1 | 839 | Missense Mutation | CTG,GTG | L,V 140 | XP_016857809.1 |