Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
CCTGTAGTGGCCCCTGTCTTGCCAG[C/T]CTGGAACACGTCTGCAGGGGACTTC
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
12 submissions
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 131241 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
EDN2 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
EDN2 - endothelin 2 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001302269.1 | 479 | Intron | NP_001289198.1 | |||
NM_001956.4 | 479 | Missense Mutation | ACT,GCT | T,A 133 | NP_001947.1 | |
XM_017000512.1 | 479 | Intron | XP_016856001.1 |