Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
CACTGGTGCTGCCCTGTACCCACAA[C/T]GTGTGCCAGGCCTGTGCCCGAGAGG
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
1 submissions
|
||||||||||||||||||||
Phenotype: |
MIM: 600986 | ||||||||||||||||||||
Literature Links: |
KRTCAP2 PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
KRTCAP2 - keratinocyte associated protein 2 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
TRIM46 - tripartite motif containing 46 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001256599.1 | 447 | Silent Mutation | AAC,AAT | N,N 28 | NP_001243528.1 | |
NM_001256600.1 | 447 | Silent Mutation | AAC,AAT | N,N 38 | NP_001243529.1 | |
NM_001256601.1 | 447 | Silent Mutation | AAC,AAT | N,N 38 | NP_001243530.1 | |
NM_001282378.1 | 447 | Intron | NP_001269307.1 | |||
NM_001282379.1 | 447 | Silent Mutation | AAC,AAT | N,N 51 | NP_001269308.1 | |
NM_025058.4 | 447 | Silent Mutation | AAC,AAT | N,N 51 | NP_079334.3 | |
XM_011510002.1 | 447 | Silent Mutation | AAC,AAT | N,N 80 | XP_011508304.1 | |
XM_017002393.1 | 447 | Silent Mutation | AAC,AAT | N,N 38 | XP_016857882.1 |