Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
GTTTGCACCTGTGCCTACTTCAAGA[A/C]AGTACCTCGTCTCAAAACCTGGCTG
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
6 submissions
|
||||||||||||||||||||
Phenotype: |
|||||||||||||||||||||
Literature Links: |
SCARNA2 PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
SCARNA2 - small Cajal body-specific RNA 2 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
TMEM167B - transmembrane protein 167B | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001322248.1 | 169 | Missense Mutation | AAA,ACA | K,T 26 | NP_001309177.1 | |
NM_020141.3 | 169 | Missense Mutation | AAA,ACA | K,T 26 | NP_064526.1 |