Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TTCGCCCGTCCCTGTCACGTCTAGC[A/G]GGCAGCCCTCCTTGTTGTGCAGGAA
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
3 submissions
|
||||||||||||||||||||
Phenotype: |
MIM: 613181 MIM: 185860 | ||||||||||||||||||||
Literature Links: |
BOLA1 PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
BOLA1 - bolA family member 1 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
SV2A - synaptic vesicle glycoprotein 2A | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001278719.1 | 2028 | Missense Mutation | CCG,CTG | P,L 36 | NP_001265648.1 | |
NM_014849.4 | 2028 | Missense Mutation | CCG,CTG | P,L 584 | NP_055664.3 | |
XM_017002992.1 | 2028 | Missense Mutation | CCG,CTG | P,L 584 | XP_016858481.1 |