Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TCCTCAACCACTGTGCTCCTGGAAC[C/T]GAAACGAAATGTCTGAAGCCCCAGC
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
3 submissions
|
||||||||||||||||||||
Phenotype: |
MIM: 609375 MIM: 609852 | ||||||||||||||||||||
Literature Links: |
LIN9 PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
LIN9 - lin-9 DREAM MuvB core complex component | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
MIXL1 - Mix paired-like homeobox | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001282402.1 | 586 | Silent Mutation | ACC,ACT | T,T 174 | NP_001269331.1 | |
NM_031944.2 | 586 | Silent Mutation | ACC,ACT | T,T 166 | NP_114150.1 |