Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
ACACCTGCCCTGGCTGTCATCTACA[C/T]GGCCAAACAGGAGCTGATGGTGGCC
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
2 submissions
|
||||||||||||||||||||
Phenotype: |
|||||||||||||||||||||
Literature Links: |
TAF5L PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
TAF5L - TATA-box binding protein associated factor 5 like | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
URB2 - URB2 ribosome biogenesis 2 homolog (S. cerevisiae) | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001314021.1 | 573 | Missense Mutation | ACG,ATG | T,M 140 | NP_001300950.1 | |
NM_014777.3 | 573 | Missense Mutation | ACG,ATG | T,M 140 | NP_055592.2 | |
XM_005273360.3 | 573 | Missense Mutation | ACG,ATG | T,M 140 | XP_005273417.1 |