Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TCTTAAATACTCACATCTTGGACTT[A/T]CTGGATGAAAGGATCTTTCCATTCA
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
2 submissions
|
||||||||||||||||||||
Phenotype: |
|||||||||||||||||||||
Literature Links: |
TMEM206 PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
TMEM206 - transmembrane protein 206 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001198862.1 | 1122 | Nonsense Mutation | AAA,TAA | K,* 355 | NP_001185791.1 | |
NM_018252.2 | 1122 | Nonsense Mutation | AAA,TAA | K,* 294 | NP_060722.2 | |
XM_005273177.3 | 1122 | Nonsense Mutation | AAA,TAA | K,* 217 | XP_005273234.1 | |
XM_011509717.2 | 1122 | Nonsense Mutation | AAA,TAA | K,* 379 | XP_011508019.1 | |
XM_011509718.2 | 1122 | Missense Mutation | AGA,AGT | R,S 330 | XP_011508020.1 | |
XM_017001634.1 | 1122 | Nonsense Mutation | AAA,TAA | K,* 309 | XP_016857123.1 | |
XM_017001635.1 | 1122 | Nonsense Mutation | AAA,TAA | K,* 224 | XP_016857124.1 | |
XM_017001636.1 | 1122 | Nonsense Mutation | AAA,TAA | K,* 217 | XP_016857125.1 | |
XM_017001637.1 | 1122 | Missense Mutation | AGA,AGT | R,S 245 | XP_016857126.1 |