Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
CAGTGTGAGTTCGCTGATGTTCAAC[A/G]AAGGAGAACTTCTGACAGAAGGTTT
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
|||||||||||||||||||||||||||||||||||||||
Literature Links: |
ZNF334 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
ZNF334 - zinc finger protein 334 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001270497.1 | 2212 | Silent Mutation | TTC,TTT | F,F 586 | NP_001257426.1 | |
NM_018102.4 | 2212 | Silent Mutation | TTC,TTT | F,F 587 | NP_060572.3 | |
XM_011528892.2 | 2212 | Silent Mutation | TTC,TTT | F,F 610 | XP_011527194.1 | |
XM_011528895.2 | 2212 | Silent Mutation | TTC,TTT | F,F 594 | XP_011527197.1 | |
XM_011528897.2 | 2212 | Silent Mutation | TTC,TTT | F,F 586 | XP_011527199.1 | |
XM_017027933.1 | 2212 | Silent Mutation | TTC,TTT | F,F 610 | XP_016883422.1 | |
XM_017027934.1 | 2212 | Silent Mutation | TTC,TTT | F,F 610 | XP_016883423.1 | |
XM_017027935.1 | 2212 | Silent Mutation | TTC,TTT | F,F 594 | XP_016883424.1 | |
XM_017027936.1 | 2212 | Silent Mutation | TTC,TTT | F,F 587 | XP_016883425.1 | |
XM_017027937.1 | 2212 | Silent Mutation | TTC,TTT | F,F 610 | XP_016883426.1 | |
XM_017027938.1 | 2212 | Silent Mutation | TTC,TTT | F,F 587 | XP_016883427.1 | |
XM_017027939.1 | 2212 | Silent Mutation | TTC,TTT | F,F 586 | XP_016883428.1 | |
XM_017027940.1 | 2212 | Silent Mutation | TTC,TTT | F,F 586 | XP_016883429.1 | |
XM_017027941.1 | 2212 | Silent Mutation | TTC,TTT | F,F 586 | XP_016883430.1 | |
XM_017027942.1 | 2212 | Silent Mutation | TTC,TTT | F,F 582 | XP_016883431.1 | |
XM_017027943.1 | 2212 | Silent Mutation | TTC,TTT | F,F 582 | XP_016883432.1 | |
XM_017027944.1 | 2212 | Silent Mutation | TTC,TTT | F,F 549 | XP_016883433.1 |